Anti-MORC2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-527
- 77845-526
- Antigen Symbol:
- MORC2
- MORC2
- Antigen Synonyms:
- AC004542.C22.1.|CW type with coiled coil domain 1|KIAA0852|MORC family CW type zinc finger 2|MORC family CW-type zinc finger protein 2|MORC2|MORC2_HUMAN|ZCW3|ZCWCC1|Zinc finger|Zinc finger CW type coiled coil domain protein 1|Zinc finger CW type with coiled coil domain 1|Zinc finger CW-type coiled-coil domain protein 1|Zing finger CW type 3
- AC004542.C22.1.|CW type with coiled coil domain 1|KIAA0852|MORC family CW type zinc finger 2|MORC family CW-type zinc finger protein 2|MORC2|MORC2_HUMAN|ZCW3|ZCWCC1|Zinc finger|Zinc finger CW type coiled coil domain protein 1|Zinc finger CW type with coiled coil domain 1|Zinc finger CW-type coiled-coil domain protein 1|Zing finger CW type 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6X9
- Q9Y6X9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human MORC family CW-type zinc finger protein 2 protein (2-300AA)
- Recombinant Human MORC family CW-type zinc finger protein 2 protein (2-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together