Anti-DRG1 Rabbit Recombinant Polyclonal Antibody
77846-451
New Product
- Antigen Symbol:DRG1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y295
- Antigen Synonyms:Developmentally regulated GTP binding protein 1|Developmentally-regulated GTP-binding protein 1|DKFZp434N1827|DRG 1|DRG-1|DRG1_HUMAN|NEDD 3|NEDD-3|NEDD3|Neural precursor cell expressed developmentally down regulated protein 3|Neural precursor cell expressed developmentally down-regulated protein 3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Developmentally-regulated GTP-binding protein 1 protein (173-228AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:200 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human