Anti-DARS Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-988
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DARS
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P14868
- Antigen Synonyms:Aspartate tRNA ligase 1 cytoplasmic|Aspartate tRNA ligase|Aspartate--tRNA ligase|Aspartyl tRNA synthetase|Aspartyl tRNA synthetase cytoplasmic|Aspartyl-tRNA synthetase|AspRS|Cell proliferation inducing gene 40 protein|Cell proliferation inducing protein 40|Cell proliferation-inducing gene 40 protein|cytoplasmic|DKFZp781B11202|MGC111579|SYDC_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aspartate--tRNA ligase, cytoplasmic protein (1-300AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DARS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human