Anti-ACIN1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: ACIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-971
- 77848-972
- Antigen Symbol:
- ACIN1
- ACIN1
- Antigen Synonyms:
- ACIN 1|ACINU_HUMAN|Acinus|ACN|Apoptotic chromatin condensation inducer 1|Apoptotic chromatin condensation inducer in the nucleus|DKFZp667N107|fSAP152|functional spliceosome-associated protein 152|KIAA0670
- ACIN 1|ACINU_HUMAN|Acinus|ACN|Apoptotic chromatin condensation inducer 1|Apoptotic chromatin condensation inducer in the nucleus|DKFZp667N107|fSAP152|functional spliceosome-associated protein 152|KIAA0670
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKV3
- Q9UKV3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ACIN1
- Synthetic peptide of Human ACIN1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together