Anti-APOOL Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77851-318
New Product
- Antigen Symbol:APOOL
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q6UXV4
- Antigen Synonyms:AAIR8193|Apolipoprotein O-like|APOOL_HUMAN|CXorf33|FAM121A|Family with sequence similarity 121A|MGC129748|Protein FAM121A|UNQ8193
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human MICOS complex subunit MIC27 protein (163-268AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: APOOL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human