Anti-CSNK1G3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: CSNK1G3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-515
- 77845-516
- Antigen Symbol:
- CSNK1G3
- CSNK1G3
- Antigen Synonyms:
- CSNK1G3|Casein kinase I isoform gamma-3|CKI-gamma 3|EC 2.7.11.1
- CSNK1G3|Casein kinase I isoform gamma-3|CKI-gamma 3|EC 2.7.11.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6M4
- Q9Y6M4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Casein kinase I isoform gamma-3 protein (1-204AA)
- Recombinant Human Casein kinase I isoform gamma-3 protein (1-204AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together