Anti-ACOX3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-651
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACOX3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O15254
- Antigen Synonyms:BRCOX|PRCOXPeroxisomal acyl-coenzyme A oxidase 3|EC 1.3.3.6|Branched-chain acyl-CoA oxidase|BRCACox|Pristanoyl-CoA oxidase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Peroxisomal acyl-coenzyme A oxidase 3 protein (401-624AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ACOX3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human