Anti-p53 (Ab-2) Mouse Monoclonal Antibody [clone: PAb181]
EMOP09-200UG
- Antibody Type:Primary
- Antigen Symbol:p53 (Ab-2)
- Clonality:Monoclonal
- Clone:PAb181
- Host:Mouse
- Isotype:IgG1
- Reactivity:Mouse,Rat,Human
- Size:200 μg
- Storage Temperature:2...8 °C
- Shipping Temperature:Wet ice
- Packaging:Plastic ampoule
Recognizes the ~53 kDa wild-type and mutant p53 protein in A-431 cells and breast carcinoma tissue.
Type: Primary
Antigen: p53 (Ab-2)
Clonality: Monoclonal
Clone: PAb181
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat;Human