Anti-MEIS2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-651
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MEIS2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:O14770
- Antigen Synonyms:Homeobox protein HsT18361|Meis (mouse) homolog 2|Meis homeobox 2|Meis homolog 2|Meis1 myeloid ecotropic viral integration site 1 homolog 2 (mouse)|Meis1 myeloid ecotropic viral integration site 1 homolog 2|Meis1 related gene 1|Meis1 related protein 1|Meis1-related protein 1|MEIS2_HUMAN|MGC2820|MRG1|TALE homeobox protein Meis2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Homeobox protein Meis2 protein (377-470AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: MEIS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse