Anti-B3GNT2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-751
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:B3GNT2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NY97
- Antigen Synonyms:3-galactosyltransferase 7|3-GalTase 7|3-Gn-T1|3-Gn-T2|3-N-acetylglucosaminyltransferase 1|3-N-acetylglucosaminyltransferase 2|b3Gal-T7|B3GN2_HUMAN|B3GNT|Beta 1 3 galactosyltransferase 7|Beta-1|Beta-3-Gx-T7|Beta3Gal-T7|Beta3GalT7|Beta3Gn T1|Beta3Gn-T1|Beta3Gn-T2|Beta3GNT|BGnT-1|BGnT-2|UDP-Gal:beta-GlcNAc beta-1|UDP-galactose:beta-N-acetylglucosamine beta-1|UDP-GlcNAc:betaGal beta-1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 2 protein (29-397AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: B3GNT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human