Anti-NME7 Rabbit Recombinant Polyclonal Antibody
77846-207
New Product
- Antigen Symbol:NME7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Antigen Synonyms:MN23H7|NDK 7|NDK7_HUMAN|NDP kinase 7|nm23 H7|nm23-H7|NME/NM23 family member 7|Nme7|Non metastatic cells 7 protein expressed in (nucleoside diphosphate kinase)|Nucleoside diphosphate kinase 7
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Nucleoside diphosphate kinase 7 protein (1-150AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: NME7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse