Anti-ETV7 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ETV7
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-625
- 77846-626
- Antigen Symbol:
- ETV7
- ETV7
- Antigen Synonyms:
- ETS related protein Tel2|Ets transcription factor TEL-2b|Ets transcription factor TEL2|ETS translocation variant 7|ets variant 7|ets variant gene 7 (TEL2 oncogene)|ets variant gene 7|ETS-related protein Tel2|ETV7_HUMAN|TEL 2|Tel related Ets factor |Tel-related Ets factor|TEL2 |TEL2 oncogene|TELB|Transcription factor Transcription factor Tel 2|Transcription factor Tel-2|TREF
- ETS related protein Tel2|Ets transcription factor TEL-2b|Ets transcription factor TEL2|ETS translocation variant 7|ets variant 7|ets variant gene 7 (TEL2 oncogene)|ets variant gene 7|ETS-related protein Tel2|ETV7_HUMAN|TEL 2|Tel related Ets factor |Tel-related Ets factor|TEL2 |TEL2 oncogene|TELB|Transcription factor Transcription factor Tel 2|Transcription factor Tel-2|TREF
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y603
- Q9Y603
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transcription factor ETV7 protein (134-218AA)
- Recombinant Human Transcription factor ETV7 protein (134-218AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together