Anti-KLK6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:100 to 1:300
Type:
Antigen: KLK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-453
- 77849-454
- Antigen Symbol:
- KLK6
- KLK6
- Antigen Synonyms:
- Bssp|hK 6|hK6|Kallikrein 6 precursor|Kallikrein related peptidase 6|Kallikrein-6|Kallikrein6|KLK 6|Klk 7|KLK 9|Klk29|KLK6_HUMAN|Klk7|KLK9|MGC9355|mGK 1|mGK1|MSP|Neurosin|Protease M|Protease serine 18|Protease serine 9|PRSS 18|PRSS 9|PRSS18|PRSS9|Serine protease 18|Serine protease 9|SP 59|SP59|Tissue kallikrein|TK|Zyme
- Bssp|hK 6|hK6|Kallikrein 6 precursor|Kallikrein related peptidase 6|Kallikrein-6|Kallikrein6|KLK 6|Klk 7|KLK 9|Klk29|KLK6_HUMAN|Klk7|KLK9|MGC9355|mGK 1|mGK1|MSP|Neurosin|Protease M|Protease serine 18|Protease serine 9|PRSS 18|PRSS 9|PRSS18|PRSS9|Serine protease 18|Serine protease 9|SP 59|SP59|Tissue kallikrein|TK|Zyme
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92876
- Q92876
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human KLK6
- Fusion protein of Human KLK6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together