Anti-PAQR5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PAQR5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-589
- 77843-588
- Antigen Symbol:
- PAQR5
- PAQR5
- Antigen Synonyms:
- MPRG|Membrane progestin receptor gamma|mPR gamma|Membrane progesterone P4 receptor gamma|Membrane progesterone receptor gamma|Progesterone and adipoQ receptor family member 5|Progestin and adipoQ receptor family member 5|Progestin and adipoQ receptor family member V
- MPRG|Membrane progestin receptor gamma|mPR gamma|Membrane progesterone P4 receptor gamma|Membrane progesterone receptor gamma|Progesterone and adipoQ receptor family member 5|Progestin and adipoQ receptor family member 5|Progestin and adipoQ receptor family member V
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NXK6
- Q9NXK6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Membrane progestin receptor gamma protein (1-51AA)
- Recombinant Human Membrane progestin receptor gamma protein (1-51AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together