Anti-GLO1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:100 to 1:300
Type:
Antigen: GLO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77848-752
- 77848-751
- Antigen Symbol:
- GLO1
- GLO1
- Antigen Synonyms:
- Aldoketomutase|GLOD1|Glx I|GLYI|glyoxalase domain containing 1|Glyoxalase I|Ketone aldehyde mutase|Ketone-aldehyde mutase|Lactoyl glutathione lyase|Lactoylglutathione lyase|LGUL_HUMAN|Methylglyoxalase|S D lactoylglutathione methylglyoxal lyase|S-D-lactoylglutathione methylglyoxal lyase
- Aldoketomutase|GLOD1|Glx I|GLYI|glyoxalase domain containing 1|Glyoxalase I|Ketone aldehyde mutase|Ketone-aldehyde mutase|Lactoyl glutathione lyase|Lactoylglutathione lyase|LGUL_HUMAN|Methylglyoxalase|S D lactoylglutathione methylglyoxal lyase|S-D-lactoylglutathione methylglyoxal lyase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q04760
- Q04760
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human GLO1
- Fusion protein of Human GLO1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together