Anti-CNBP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77853-312
New Product
- Antigen Symbol:CNBP
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P62633
- Antigen Synonyms:CCHC type zinc finger nucleic acid binding protein|Cellular nucleic acid binding protein|Cellular nucleic acid-binding protein|CNBP|CNBP_HUMAN|CNBP1|DM2|Erythroid differentiation related|PROMM|Proximal myotonic myopathy nucleic acid binding protein|RNF163|Sterol regulatory element binding protein|ZCCHC22|Zinc finger protein 273|Zinc finger protein 9 (a cellular retroviral nucleic acid binding protein)|Zinc finger protein 9|ZNF9
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cellular nucleic acid-binding protein (2-170AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Type:
Antigen: CNBP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human