Anti-CTAGE5 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77853-707
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CTAGE5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O15320
- Antigen Synonyms:Endoplasmic reticulum export factor CTAGE5 (CTAGE family member 5 ER export factor) (Protein cTAGE-5) (Meningioma-expressed antigen 6/11), CTAGE5, MEA11 MEA6 MGEA11 MGEA6
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Endoplasmic reticulum export factor CTAGE5 protein (515-632AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: CTAGE5
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human