Anti-NEDD4L Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: NEDD4L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77849-432
- 77849-433
- Antigen Symbol:
- NEDD4L
- NEDD4L
- Antigen Synonyms:
- E3 ubiquitin protein ligase NEDD4 like protein|E3 ubiquitin-protein ligase NEDD4-like|KIAA0439|NED4L_HUMAN|NEDD4 2|NEDD4 2c|Nedd4-2|NEDD4-2a|NEDD4.2|NEDD4b|NEDD4L|NEDD4La|NEDD4Lb|NEDD4Lc|NEDD4Ld|NEDD4Le|NEDD4Lf|NEDD4Lg|NEDD4Lh|NEDL3|Neural precursor cell expressed developmentally down regulated 4 like E3 ubiquitin protein ligase|Neural precursor cell expressed|developmentally down regulated 4 like|RSP5|Ubiquitin protein ligase Rsp5
- E3 ubiquitin protein ligase NEDD4 like protein|E3 ubiquitin-protein ligase NEDD4-like|KIAA0439|NED4L_HUMAN|NEDD4 2|NEDD4 2c|Nedd4-2|NEDD4-2a|NEDD4.2|NEDD4b|NEDD4L|NEDD4La|NEDD4Lb|NEDD4Lc|NEDD4Ld|NEDD4Le|NEDD4Lf|NEDD4Lg|NEDD4Lh|NEDL3|Neural precursor cell expressed developmentally down regulated 4 like E3 ubiquitin protein ligase|Neural precursor cell expressed|developmentally down regulated 4 like|RSP5|Ubiquitin protein ligase Rsp5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96PU5
- Q96PU5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human NEDD4L
- Fusion protein of Human NEDD4L
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together