Anti-CLINT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77853-256
New Product
- Antigen Symbol:INSR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P06213
- Antigen Synonyms:CD220|HHF5|human insulin receptor|Insr|INSR_HUMAN|Insulin receptor subunit beta|IR 1|IR|IR-1 |IR1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:CD220
- Purification:Antigen affinity purified
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human