Anti-CRNN Rabbit Recombinant polyclonal Antibody (Biotinylated)
Catalog # 77853-587
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRNN
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UBG3
- Antigen Synonyms:53 kDa putative calcium binding protein|53 kDa putative calcium-binding protein|53 kDa squamous epithelial induced stress protein|53 kDa squamous epithelial-induced stress protein|58 kDa heat shock protein|C1orf10|Chromosome 1 open reading frame 10|Cornulin|CRNN_HUMAN|DRC1|PDRC1|SEP53|Squamous epithelial heat shock protein 53|Tumor related protein|Tumor-related protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Cornulin protein (139-389AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: CRNN
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human