Anti-ATP5F1D Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-618
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP5F1D
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P30049
- Antigen Synonyms:ATP synthase subunit delta|mitochondrial|ATP synthase|H+ transporting|mitochondrial F1 complex|delta subunit|ATP5D|ATPD_HUMAN|F ATPase delta subunit|F-ATPase delta subunit|Mitochondrial ATP synthase complex delta subunit precusor|Mitochondrial ATP synthase delta subunit
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP synthase subunit delta, mitochondrial protein (23-168AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human