Anti-FZD10 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FZD10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-493
- 77844-492
- Antigen Symbol:
- FZD10
- FZD10
- Antigen Synonyms:
- CD 350|CD 350 antigen|CD350|Frizzled (Drosophila) homolog 10|Frizzled-10|FZ 10|Fz-10|FZ10|FZD 10|FZD10_HUMAN|FzE 7|FzE7|hFz 10|hFz10
- CD 350|CD 350 antigen|CD350|Frizzled (Drosophila) homolog 10|Frizzled-10|FZ 10|Fz-10|FZ10|FZD 10|FZD10_HUMAN|FzE 7|FzE7|hFz 10|hFz10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULW2
- Q9ULW2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Frizzled-10 protein (125-225AA)
- Recombinant Human Frizzled-10 protein (125-225AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together