Anti-USE1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: USE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-623
- 77843-622
- Antigen Symbol:
- USE1
- USE1
- Antigen Synonyms:
- 2010315L10Rik|5730403H22Rik|AV002165|D12|Ed2|Embryonic development factor 2|MDS032|P31|Protein D12|Protein p31|Putative MAPK activating protein PM26|Putative MAPK-activating protein PM26|Q-snare|RGD1306660|SLT1|SNARE-like tail-anchored protein 1 homolog|Unconventional SNARE in the ER 1 homolog (S. cerevisiae)|Unconventional SNARE in the ER 1 homolog|USE1 like protein|Use1 unconventional SNARE in the ER 1 homolog (S. cerevisiae)|USE1-like protein|USE1_HUMAN|USE1L|Vesicle transport protein USE1
- 2010315L10Rik|5730403H22Rik|AV002165|D12|Ed2|Embryonic development factor 2|MDS032|P31|Protein D12|Protein p31|Putative MAPK activating protein PM26|Putative MAPK-activating protein PM26|Q-snare|RGD1306660|SLT1|SNARE-like tail-anchored protein 1 homolog|Unconventional SNARE in the ER 1 homolog (S. cerevisiae)|Unconventional SNARE in the ER 1 homolog|USE1 like protein|Use1 unconventional SNARE in the ER 1 homolog (S. cerevisiae)|USE1-like protein|USE1_HUMAN|USE1L|Vesicle transport protein USE1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZ43
- Q9NZ43
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Vesicle transport protein USE1 protein (1-146AA)
- Recombinant human Vesicle transport protein USE1 protein (1-146AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together