Anti-MGST2 Rabbit Polyclonal Antibody
Catalog # 77850-038
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MGST2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q99735
- Antigen Synonyms:FLJ27438|glutathione S transferase|microsomal|2|GST 2|GST2|MGC14097|MGST 2|MGST II|MGST2_HUMAN|MGSTII|Microsomal glutathione S transferase 2|Microsomal glutathione S-transferase 2|Microsomal GST 2|Microsomal GST II
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Internal of Human MGST2.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:3000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human