Anti-LAS1L Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LAS1L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-198
- 77846-199
- Antigen Symbol:
- LAS1L
- LAS1L
- Antigen Synonyms:
- dJ475B7.2|FLJ12525|FLJ30133|FLJ45274|LAS1 like (S. cerevisiae)|Las1 like|LAS1L_HUMAN|Protein LAS1 homolog|Ribosomal biogenesis protein RP3 475B7.2
- dJ475B7.2|FLJ12525|FLJ30133|FLJ45274|LAS1 like (S. cerevisiae)|Las1 like|LAS1L_HUMAN|Protein LAS1 homolog|Ribosomal biogenesis protein RP3 475B7.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4W2
- Q9Y4W2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ribosomal biogenesis protein LAS1L protein (490-565AA)
- Recombinant Human Ribosomal biogenesis protein LAS1L protein (490-565AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together