Anti-HES7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HES7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-988
- 77842-987
- Antigen Symbol:
- HES7
- HES7
- Antigen Synonyms:
- bHLH factor bHLHb37|Class B basic helix loop helix protein 37|Class B basic helix-loop-helix protein 37|Hairy and enhancer of split 7|hes family bHLH transcription factor 7|HES7_HUMAN|hSCDO4|Transcription factor HES 7|Transcription factor HES-7
- bHLH factor bHLHb37|Class B basic helix loop helix protein 37|Class B basic helix-loop-helix protein 37|Hairy and enhancer of split 7|hes family bHLH transcription factor 7|HES7_HUMAN|hSCDO4|Transcription factor HES 7|Transcription factor HES-7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BYE0
- Q9BYE0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transcription factor HES-7 protein (127-192AA)
- Recombinant Human Transcription factor HES-7 protein (127-192AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together