Anti-MBNL1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MBNL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-427
- 77843-426
- Antigen Symbol:
- MBNL1
- MBNL1
- Antigen Synonyms:
- EXP|EXP35|EXP40|EXP42|KIAA0428|MBNL|MBNL protein|MBNL1_HUMAN|Muscleblind 41kD isoform|Muscleblind like|Muscleblind like protein 1|Muscleblind like splicing regulator 1|Muscleblind-like protein 1|Triplet expansion RNA binding protein|Triplet-expansion RNA-binding protein
- EXP|EXP35|EXP40|EXP42|KIAA0428|MBNL|MBNL protein|MBNL1_HUMAN|Muscleblind 41kD isoform|Muscleblind like|Muscleblind like protein 1|Muscleblind like splicing regulator 1|Muscleblind-like protein 1|Triplet expansion RNA binding protein|Triplet-expansion RNA-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NR56
- Q9NR56
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Muscleblind-like protein 1 protein (1-382AA)
- Recombinant Human Muscleblind-like protein 1 protein (1-382AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together