Anti-EGLN1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:10 to 1:50
Type:
Antigen: EGLN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77848-767
- 77848-768
- Antigen Symbol:
- EGLN1
- EGLN1
- Antigen Synonyms:
- C1ORF12|Chromosome 1 Open Reading Frame 12|DKFZp761F179|ECYT 3|ECYT3|Egl 9 family hypoxia inducible factor 1|EGL 9 homolog of C. elegans 1|EGL nine (C.elegans) homolog 1|Egl nine homolog 1 (C. elegans)|Egl nine homolog 1|Egl nine like protein 1|EGLN 1|EGLN1_HUMAN|HIF PH2|HIF Prolyl Hydroxylase 2|HIF-PH2|HIF-prolyl hydroxylase 2|HIFP4H 2|HIFPH2|HPH 2|HPH-2|HPH2|Hypoxia inducible factor prolyl hydroxylase 2|Hypoxia-inducible factor prolyl hydroxylase 2|ORF13|P4H2|PHD 2|PhD2|PNAS 118|PNAS 137|Prolyl Hydroxylase Domain Containing Protein 2|Prolyl hydroxylase domain-containing protein 2|Rat Homolog of SM20|SM 20|SM-20|SM20|Zinc finger MYND domain containing protein 6|ZMYND6
- C1ORF12|Chromosome 1 Open Reading Frame 12|DKFZp761F179|ECYT 3|ECYT3|Egl 9 family hypoxia inducible factor 1|EGL 9 homolog of C. elegans 1|EGL nine (C.elegans) homolog 1|Egl nine homolog 1 (C. elegans)|Egl nine homolog 1|Egl nine like protein 1|EGLN 1|EGLN1_HUMAN|HIF PH2|HIF Prolyl Hydroxylase 2|HIF-PH2|HIF-prolyl hydroxylase 2|HIFP4H 2|HIFPH2|HPH 2|HPH-2|HPH2|Hypoxia inducible factor prolyl hydroxylase 2|Hypoxia-inducible factor prolyl hydroxylase 2|ORF13|P4H2|PHD 2|PhD2|PNAS 118|PNAS 137|Prolyl Hydroxylase Domain Containing Protein 2|Prolyl hydroxylase domain-containing protein 2|Rat Homolog of SM20|SM 20|SM-20|SM20|Zinc finger MYND domain containing protein 6|ZMYND6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZT9
- Q9GZT9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human EGLN1
- Fusion protein of Human EGLN1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together