Anti-HS2ST1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-657
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HS2ST1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q7LGA3
- Antigen Synonyms:2-O-sulfotransferase|2OST|dJ604K5.2|FLJ11317|heparan sulfate 2 O sulfotransferase 1|Heparan sulfate 2-O-sulfotransferase 1|HS2ST_HUMAN|KIAA0448|MGC131986
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Heparan sulfate 2-O-sulfotransferase 1 protein (29-229AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: HS2ST1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat