Anti-CAPN1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-331
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CAPN1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:P07384
- Antigen Synonyms:Ca2 activated neutral protease|Calcium activated neutral proteinase|Calcium activated neutral proteinase small subunit|Calcium dependent protease small subunit 1|Calcium dependent protease small subunit|Calcium-activated neutral proteinase 1|Calpain 1|Calpain 1 large subunit|(mu/I) large subunit|Calpain mu type|Calpain mu-type|Calpain regulatory subunit|Calpain small subunit 1|Calpain|large polypeptide L1|Calpain-1 catalytic subunit|Calpain-1 large subunit|CAN1_HUMAN|CANP 1|CANP|CANP small subunit|CANP1|CANPL 1|CANPL1|CAPN 1|Cell proliferation inducing protein 30|Cell proliferation-inducing gene 30 protein|Micromolar Calpain|Micromolar-calpain|Mu Calpain|muCANP|muCL
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Calpain-1 catalytic subunit protein (20-330AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: CAPN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human