Anti-CCNH Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77852-529
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CCNH
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P51946
- Antigen Synonyms:6330408H09Rik|AI661354|AV102684|AW538719|CAK|CAK complex subunit|ccnh|CCNH_HUMAN|CDK activating kinase|CDK activating kinase complex subunit|Cyclin dependent kinase activating kinase|cyclin dependent kinase activating kinase complex subunit|Cyclin H|Cyclin-H|CyclinH|MO15 associated protein|MO15-associated protein|p34|p36|p37
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cyclin-H protein (261-319AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CCNH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human