Anti-IVNS1ABP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: IVNS1ABP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-042
- 77846-043
- Antigen Symbol:
- IVNS1ABP
- IVNS1ABP
- Antigen Synonyms:
- ARA3|FLARA3|KIAA0850|KLHL39|NS1|NS1BP|HSPC068|Influenza virus NS1A-binding protein|NS1-BP|NS1-binding protein|Aryl hydrocarbon receptor-associated protein 3|Kelch-like protein 39
- ARA3|FLARA3|KIAA0850|KLHL39|NS1|NS1BP|HSPC068|Influenza virus NS1A-binding protein|NS1-BP|NS1-binding protein|Aryl hydrocarbon receptor-associated protein 3|Kelch-like protein 39
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6Y0
- Q9Y6Y0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Influenza virus NS1A-binding protein (401-642AA)
- Recombinant Human Influenza virus NS1A-binding protein (401-642AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together