Anti-AP2M1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-840
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AP2M1
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q96CW1
- Antigen Synonyms:Adapter-related protein complex 2 mu subunit| Adaptin mu 1| Adaptin-mu2| Adaptor protein complex AP 2 subunit mu| Adaptor protein complex AP-2 subunit mu| Adaptor related protein complex 2 mu 1 subunit| AP 2 mu 2 chain| AP-2 complex subunit mu| AP-2 mu chain| Ap2m1| AP2M1_HUMAN| AP50| CLAPM1| Clathrin adaptor complex AP2 mu subunit| Clathrin assembly protein complex 2 medium chain| Clathrin associated/assembly/adaptor protein medium 1| Clathrin coat adaptor protein AP50| Clathrin coat assembly protein AP50| Clathrin coat associated protein AP50| Clathrin coat-associated protein AP50| HA2 50 kDa subunit| mu2| Plasma membrane adaptor AP-2 50 kDa protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-2 complex subunit mu protein (1-435AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: AP2M1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human