Anti-SCN11A Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: SCN11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77847-633
- 77847-632
- Antigen Symbol:
- SCN11A
- SCN11A
- Antigen Synonyms:
- hNaN|NaN|NAV1.9|Peripheral nerve sodium channel 5|PN 5|PN5|SCN 11A|SCN 12A|Scn11a|SCN12A|SCNBA_HUMAN|Sensory neuron sodium channel 2|SNS 2|SNS2|Sodium channel protein type 11 subunit alpha|Sodium channel protein type XI subunit alpha|Sodium channel voltage gated type XI alpha|Sodium channel voltage gated type XI alpha polypeptide|Sodium channel voltage gated type XI alpha subunit|Sodium channel voltage gated type XII alpha polypeptide|Voltage gated sodium channel Nav1.9|Voltage gated sodium channel subunit alpha Nav1.9|Voltage-gated sodium channel subunit alpha Nav1.9
- hNaN|NaN|NAV1.9|Peripheral nerve sodium channel 5|PN 5|PN5|SCN 11A|SCN 12A|Scn11a|SCN12A|SCNBA_HUMAN|Sensory neuron sodium channel 2|SNS 2|SNS2|Sodium channel protein type 11 subunit alpha|Sodium channel protein type XI subunit alpha|Sodium channel voltage gated type XI alpha|Sodium channel voltage gated type XI alpha polypeptide|Sodium channel voltage gated type XI alpha subunit|Sodium channel voltage gated type XII alpha polypeptide|Voltage gated sodium channel Nav1.9|Voltage gated sodium channel subunit alpha Nav1.9|Voltage-gated sodium channel subunit alpha Nav1.9
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UI33
- Q9UI33
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SCN11A
- Synthetic peptide of Human SCN11A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together