Anti-TGM5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: TGM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77847-274
- 77847-273
- Antigen Symbol:
- TGM5
- TGM5
- Antigen Synonyms:
- Protein glutamine gamma glutamyltransferase 5|Protein-glutamine gamma-glutamyltransferase 5|PSS2|TG(X)|TGase 5|TGase X|TGase-5|TGase5|TGASEX|TGM5_HUMAN|TGM6|TGMX|TGX|Transglutaminase X|Transglutaminase-5
- Protein glutamine gamma glutamyltransferase 5|Protein-glutamine gamma-glutamyltransferase 5|PSS2|TG(X)|TGase 5|TGase X|TGase-5|TGase5|TGASEX|TGM5_HUMAN|TGM6|TGMX|TGX|Transglutaminase X|Transglutaminase-5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43548
- O43548
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TGM5
- Fusion protein of Human TGM5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together