Anti-APIP Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:500 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: APIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77847-511
- 77847-510
- Antigen Symbol:
- APIP
- APIP
- Antigen Synonyms:
- Apaf1 Interacting Protein|APAF1-interacting protein|APIP2|CG129|CGI 29|CGI29|dJ179L10.2|MMRP19|Monocyte/macrophage protein 19|MTNB_HUMAN|MTRu 1 P dehydratase|MTRu-1-P dehydratase|Probable methylthioribulose 1 phosphate dehydratase|Probable methylthioribulose-1-phosphate dehydratase
- Apaf1 Interacting Protein|APAF1-interacting protein|APIP2|CG129|CGI 29|CGI29|dJ179L10.2|MMRP19|Monocyte/macrophage protein 19|MTNB_HUMAN|MTRu 1 P dehydratase|MTRu-1-P dehydratase|Probable methylthioribulose 1 phosphate dehydratase|Probable methylthioribulose-1-phosphate dehydratase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GX9
- Q96GX9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human APIP
- Fusion protein of Human APIP
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together