Anti-CNN3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77853-329
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CNN3
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q15417
- Antigen Synonyms:Acidic calponin 3|acidic isoform|Calponin 3 acidic|Calponin 3|Calponin acidic|Calponin acidic isoform|Calponin|Calponin-3|Calponin3|CNN 3|CNN3_HUMAN|dJ639P13.2.2|OTTHUMP00000012470|OTTHUMP00000012471
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Calponin-3 protein (2-329AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CNN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human