Anti-ALDH3A1 Rabbit Recombinant polyclonal Antibody
Catalog # 77851-037
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ALDH3A1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P30838
- Antigen Synonyms:Acetaldehyde dehydrogenase 3|AHD 4|AHD C|AHD4|AHDC|AL3A1_HUMAN|Aldehyde dehydrogenase 3|Aldehyde dehydrogenase 3 family member A1|Aldehyde dehydrogenase 3A1|Aldehyde dehydrogenase|Aldehyde dehydrogenase class 3|Aldehyde dehydrogenase dimeric NADP preferring|Aldehyde dehydrogenase family 3 member A1|Aldehyde dehydrogenase family 3 subfamily A1|Aldehyde dehydrogenase family 3, subfamily A, member 1|Aldehyde dehydrogenase isozyme 3|Aldehyde dehydrogenase type III|ALDH 3|ALDH 3A1|ALDH III|ALDH, stomach type|ALDH3 A1|ALDH3|ALDH3A 1|ALDHIII|dimeric NADP-preferring|MGC10406|Stomach aldehyde dehydrogenase|Tumor associated aldehyde dehydrogenase tumor ALDH
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Aldehyde dehydrogenase, dimeric NADP-preferring protein (210-453AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:200-1:2000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: ALDH3A1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human