Anti-DNAJB2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77854-252
New Product
- Antigen Symbol:DNAJB2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P25686
- Antigen Synonyms:HSJ1|HSPF3DnaJ homolog subfamily B member 2|Heat shock 40 kDa protein 3|Heat shock protein J1|HSJ-1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DnaJ homolog subfamily B member 2 protein (2-324AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: DNAJB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human