Anti-COX5A Rabbit Polyclonal Antibody
Catalog # 77849-783
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:COX5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P20674
- Antigen Synonyms:COX|COX VA|COX5A_HUMAN|Cytochrome c oxidase polypeptide Va|Cytochrome c oxidase polypeptide|mitochondrial|Cytochrome c oxidase subunit 5A|Cytochrome c oxidase subunit Va|Mitochondrial cytochrome c oxidase subunit Va|VA
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Internal of Human COX5A.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:3000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human