Anti-UTP11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UTP11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-529
- 77846-530
- Antigen Symbol:
- UTP11
- UTP11
- Antigen Synonyms:
- CGI 94|CGI94|Probable U3 small nucleolar RNA associated protein 11|Probable U3 small nucleolar RNA-associated protein 11|U3 snoRNA associated protein 11|U3 snoRNA-associated protein 11|UTP 11L|UTP11 like protein|UTP11 like U3 small nucleolar ribonucleoprotein|UTP11-like protein|UTP11_HUMAN|Utp11l
- CGI 94|CGI94|Probable U3 small nucleolar RNA associated protein 11|Probable U3 small nucleolar RNA-associated protein 11|U3 snoRNA associated protein 11|U3 snoRNA-associated protein 11|UTP 11L|UTP11 like protein|UTP11 like U3 small nucleolar ribonucleoprotein|UTP11-like protein|UTP11_HUMAN|Utp11l
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3A2
- Q9Y3A2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable U3 small nucleolar RNA-associated protein 11 protein (1-253AA)
- Recombinant Human Probable U3 small nucleolar RNA-associated protein 11 protein (1-253AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together