Anti-COL4A5 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Catalog # 77853-392
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:COL4A5
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P29400
- Antigen Synonyms:CO4A5_HUMAN|Collagen alpha-5(IV) chain
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Collagen alpha-5(IV) chain protein (1184-1235AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: COL4A5
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human