α-Melanocyte-Stimulating Hormone
Catalog # 80603-770
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:581-05-5
- Size:1 mg
- Protein Synonyms:α-MSH|Melanotropin|α-Melanocyte-Stimulating Hormone
- Protein/Peptide Name:α-Melanocyte-Stimulating Hormone
- Purity:≥95% (HPLC)
Specifications
About this item
α-Melanocyte-Stimulating Hormone, Synthetic α-melanocyte-stimulating hormone.
Synthetic α-melanocyte-stimulating hormone. Peptide hormone secreted by the anterior pituitary gland. Controls the intensity of pigmentation. A potent inhibitor of ICAM-1 expression in malignant melanocytes.