Anti-ADAM29 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:50 to 1:200
Type:
Antigen: ADAM29
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-423
- 77844-422
- Antigen Symbol:
- ADAM29
- ADAM29
- Antigen Synonyms:
- A disintegrin and metalloproteinase domain 29|ADA29_HUMAN|ADAM 29|ADAM metallopeptidase domain 29|Cancer/testis antigen 73|CT73|Disintegrin and metalloproteinase domain-containing protein 29|Metallaproteinase disintegrin (ADAM29)|svph1
- A disintegrin and metalloproteinase domain 29|ADA29_HUMAN|ADAM 29|ADAM metallopeptidase domain 29|Cancer/testis antigen 73|CT73|Disintegrin and metalloproteinase domain-containing protein 29|Metallaproteinase disintegrin (ADAM29)|svph1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKF5
- Q9UKF5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Disintegrin and metalloproteinase domain-containing protein 29 protein (701-820AA)
- Recombinant Human Disintegrin and metalloproteinase domain-containing protein 29 protein (701-820AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together