Anti-COX5A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77852-159
New Product
- Antigen Symbol:COX5A
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P20674
- Antigen Synonyms:COX|COX VA|COX5A_HUMAN|Cytochrome c oxidase polypeptide Va|Cytochrome c oxidase polypeptide|mitochondrial|Cytochrome c oxidase subunit 5A|Cytochrome c oxidase subunit Va|Mitochondrial cytochrome c oxidase subunit Va|VA
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cytochrome c oxidase subunit 5A, mitochondrial protein (42-150AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: COX5A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human