Anti-CRIP1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-558
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRIP1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P50238
- Antigen Synonyms:CRHP|CRIP|CRIP1_HUMAN|CRP-1|CRP1|Cysteine rich heart protein|Cysteine rich intestinal protein|Cysteine rich protein 1 (intestinal)|Cysteine rich protein 1|Cysteine-rich heart protein|Cysteine-rich intestinal protein|Cysteine-rich protein 1|FLJ40971|hCRHP
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cysteine-rich protein 1 protein (1-77AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: CRIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human