Anti-APOB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-289
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:APOB
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P04114
- Antigen Synonyms:Apo B 100|Apo B|Apo B-100|Apo B-48|Apo B100|Apo B48|ApoB 100|ApoB 48|APOB_HUMAN|Apolipoprotein B (including Ag(x) antigen)|Apolipoprotein B 100|Apolipoprotein B 48|Apolipoprotein B|Apolipoprotein B-48|Apolipoprotein B100|Apolipoprotein B48|FLDB|LDLCQ4
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Apolipoprotein B-100 protein (28-127AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: APOB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human