Anti-TMED3 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: TMED3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-156
- 77846-157
- Antigen Symbol:
- TMED3
- TMED3
- Antigen Synonyms:
- C15orf22|Integral type I protein|Membrane protein p24B|p24 family protein gamma 4|p24 family protein gamma-4|P24B|p24g4|p24gamma4|p26|Testis tissue sperm binding protein Li 48e|TMED3_HUMAN|Transmembrane emp24 domain containing protein 3|Transmembrane emp24 domain-containing protein 3|Transmembrane p24 trafficking protein 3|UNQ5357/PRO1078
- C15orf22|Integral type I protein|Membrane protein p24B|p24 family protein gamma 4|p24 family protein gamma-4|P24B|p24g4|p24gamma4|p26|Testis tissue sperm binding protein Li 48e|TMED3_HUMAN|Transmembrane emp24 domain containing protein 3|Transmembrane emp24 domain-containing protein 3|Transmembrane p24 trafficking protein 3|UNQ5357/PRO1078
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3Q3
- Q9Y3Q3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Transmembrane emp24 domain-containing protein 3 protein (24-180AA)
- Recombinant human Transmembrane emp24 domain-containing protein 3 protein (24-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together