Anti-BDKRB2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77851-854
New Product
- Antigen Symbol:BDKRB2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P30411
- Antigen Synonyms:B2|B2 bradykinin receptor|B2BKR|B2BRA|B2R|BDKR B2|BDKRB 2|BDKRB2|BK 2|BK 2 receptor|BK R2|BK-2 receptor|BK2|BK2 receptor|BK2R|BKR 2|BKR2|BKRB2_HUMAN|BR B2|Bradykinin receptor B2|Bradykinin receptor beta 2|BRB 2|BRB2|DKFZp686O088|Kinin B2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human B2 bradykinin receptor protein (336-391AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: BDKRB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human