Anti-SLCO1B1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLCO1B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-026
- 77846-027
- Antigen Symbol:
- SLCO1B1
- SLCO1B1
- Antigen Synonyms:
- Liver-specific organic anion transporter 1|LST-1|LST1|OATP-2|OATP-C|OATP1B1|OATPC|SLC21A6|SO1B1_HUMAN|Sodium-independent organic anion-transporting polypeptide 2|Solute carrier family 21 member 6|Solute carrier organic anion transporter family member 1B1
- Liver-specific organic anion transporter 1|LST-1|LST1|OATP-2|OATP-C|OATP1B1|OATPC|SLC21A6|SO1B1_HUMAN|Sodium-independent organic anion-transporting polypeptide 2|Solute carrier family 21 member 6|Solute carrier organic anion transporter family member 1B1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6L6
- Q9Y6L6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Solute carrier organic anion transporter family member 1B1 protein (426-537AA)
- Recombinant Human Solute carrier organic anion transporter family member 1B1 protein (426-537AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together